Learn More
Trajan Scientific HPLC Reverse Phase Protecol Polar 3/PK
SDP
Catalog No. 5000493855
Encompass_Preferred
C18P x 3 m particle size x 120 pore size 10 mm length x 4 0 mm ID guard ProteCol HPLC column PK3
Catalog No. 50-004-93855 Supplier Trajan Scientific Supplier No. 2POL03G04
C18P x 3 m particle size x 120 pore size 10 mm length x 4 0 mm ID guard ProteCol HPLC column PK3