Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPLC Solvent Kit, HPLC, J.T. Baker™

Catalog No. 02003152 Shop All Avantor Products
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-152 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-152 Supplier Avantor Supplier No. 964500

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry

Specifications

Chemical Name or Material HPLC Solvent Kit
Grade HPLC
Quantity 6 x 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.