Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Standards Kit, UV, HPLC, Uracil, Caffeine, 10 Ampules

Catalog No. 50792905 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit provides standards for assessing Alliance HPLC systems equipped with a UV detector. It includes 10 ampules of standard solutions containing varying concentrations of Uracil and Caffeine for system assessment.

  • 10 ampules, 10 mL standard solutions each.
  • Solutions 1-5 contain 0.030 mg/mL Uracil ±1% with varying Caffeine concentrations (0.0100-0.2200 mg/mL, ±0.5%).
  • Solution 6: 0.010 mg/mL Uracil ±1%.
  • Solution 7: 0.40 μg/mL Caffeine ±0.5%.
  • Solution 8: 4.00 mg/mL Caffeine ±0.5%.
  • Solution 9: Blank mobile phase.
  • Caffeine and Uracil purity > 99%.
  • Standards in 30% HPLC Grade Methanol and 70% 18M ASTM Type 1 Lab Water, V/V.
  • Certificate of Analysis available.

Catalog No. 50-792-905 Supplier Waters Corp Supplier No. 700003741
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit provides standards for assessing Alliance HPLC systems equipped with a UV detector. It includes 10 ampules of standard solutions containing varying concentrations of Uracil and Caffeine for system assessment.

  • 10 ampules, 10 mL standard solutions each.
  • Solutions 1-5 contain 0.030 mg/mL Uracil ±1% with varying Caffeine concentrations (0.0100-0.2200 mg/mL, ±0.5%).
  • Solution 6: 0.010 mg/mL Uracil ±1%.
  • Solution 7: 0.40 μg/mL Caffeine ±0.5%.
  • Solution 8: 4.00 mg/mL Caffeine ±0.5%.
  • Solution 9: Blank mobile phase.
  • Caffeine and Uracil purity > 99%.
  • Standards in 30% HPLC Grade Methanol and 70% 18M ASTM Type 1 Lab Water, V/V.
  • Certificate of Analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.