Learn More
Agilent Technologies HPLC TUBING 116 X010 X 5 FT
Catalog No. NC2192417
Encompass
Tubing, PEEK, 1.6 mm od, 0.25 mm id, 1.5 m10 per pk
This item is not returnable. View return policy
Catalog No. NC2192417 Supplier Agilent Technologies Supplier No. 08901762
Tubing, PEEK, 1.6 mm od, 0.25 mm id, 1.5 m10 per pk