Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 5 µm, 2.1 x 30 mm HPLC Column
SDP
Catalog No. 5000150903
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-50903 Supplier Mac-Mod Supplier No. MMHB821035
Add to Cart
Add to Cart
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.