Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 5 µm, 10.0 x 100 mm HPLC Column
SDP
Catalog No. 5000151823
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-51823 Supplier Mac-Mod Supplier No. MMHB810255
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.