Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 5 µm, 3.0 x 250 mm HPLC Column
SDP
Catalog No. 5000150869
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-50869 Supplier Mac-Mod Supplier No. MMHB830255
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.