Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 3 µm, 4.6 x 30 mm HPLC Column
SDP
Catalog No. 5000150993
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-50993 Supplier Mac-Mod Supplier No. MMHB846033
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.