Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 3 µm, 4.6 x 150 mm HPLC Column
SDP
Catalog No. 5000151739
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-51739 Supplier Mac-Mod Supplier No. MMHB846153
Add to Cart
Add to Cart
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.