Learn More
Mac-Mod MonoSphere C18/SCX-M, 120 Å, 5 µm, 4.6 x 150 mm Monodisperse Fully Porous Particle HPLC Column
SDP
Catalog No. 5000151116
Encompass
Monosphere HPLC columns are designed for unique applications - in this case the C18/SCX-M is used for the analysis of Tween 80 a common surfactant used in drug product formulations
Catalog No. 50-001-51116 Supplier Mac-Mod Supplier No. MSIEX18X124615
Monosphere HPLC columns are designed for unique applications - in this case the C18/SCX-M is used for the analysis of Tween 80 a common surfactant used in drug product formulations