Learn More
Mac-Mod MAC-MOD Monodisperse SEC Diol, 500 Å, 1.8 µm, 4.6 x 300 mm LC Column
SDP
Catalog No. 5000151994
Encompass
MAC-Mod Monodisperse Particle Technology is manufactured with next generation Monodisperse Particle technology, and is avaialble in a rnage of bonded phases, pore sizes and particle sizes.
Catalog No. 50-001-51994 Supplier Mac-Mod Supplier No. MMZD3183046
MAC-Mod Monodisperse Particle Technology is manufactured with next generation Monodisperse Particle technology, and is avaialble in a rnage of bonded phases, pore sizes and particle sizes.