Learn More
Mac-Mod Avantor Partisil Silica, 85 Å, 10 µm, 4.6 x 250 mm HPLC Column
SDP
Catalog No. 5000151957
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-51957 Supplier Mac-Mod Supplier No. P10250A
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.