Learn More
Mac-Mod ProntoSIL Amino E, 120 Å, 5 µm, 2.0 x 150 mm HPLC Column
SDP
Catalog No. 5000144391
Encompass
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.
Catalog No. 50-001-44391 Supplier Mac-Mod Supplier No. 1502F190PS050E
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.