Learn More
Mac-Mod Avantor Prevail CN, 110 Å, 3 µm, 4.6 x 150 mm HPLC Column
SDP
Catalog No. 5000152577
Encompass
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.
Catalog No. 50-001-52577 Supplier Mac-Mod Supplier No. 99247 / 5131357
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.