Learn More
Mac-Mod ProntoSIL C30, 200 Å, 10 µm, 50.0 x 250 mm HPLC Column
SDP
Catalog No. 5000144770
Encompass
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.
Catalog No. 50-001-44770 Supplier Mac-Mod Supplier No. 2550H300PS100
Add to Cart
Add to Cart
Request a Quote
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.