Learn More
Mac-Mod ProntoSIL Amino E, 120 Å, 5 µm, 2.0 x 30 mm HPLC Column
SDP
Catalog No. 5000145830
Encompass
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.
Catalog No. 50-001-45830 Supplier Mac-Mod Supplier No. 0302F190PS050E
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.