Learn More
Mac-Mod ProntoSIL C8 SH, 120 Å, 3 µm, 4.6 x 33 mm HPLC Column
SDP
Catalog No. 5000145654
Encompass
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.
Catalog No. 50-001-45654 Supplier Mac-Mod Supplier No. 0446F080PS030
Add to Cart
Add to Cart
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.