Learn More
Mac-Mod MONOBIO HIC Butyl 1000 Å, 5 µm, 4.6 x 50 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152241
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-52241 Supplier Mac-Mod Supplier No. MBHICHBU424605
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)