Learn More
Mac-Mod MONOBIO HIC Butyl 1000 Å, 3 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152331
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-52331 Supplier Mac-Mod Supplier No. MBHICHBU414610
Add to Cart
Add to Cart
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)