Learn More
Mac-Mod MONOBIO HIC-OH 1000 Å, 3 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152330
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-52330 Supplier Mac-Mod Supplier No. MBHICHPG414610
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)