Learn More
Mac-Mod MONOBIO HIC-Phenyl 1000 Å, 3 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000151727
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-51727 Supplier Mac-Mod Supplier No. MBHICHPH414610
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)