Learn More
Mac-Mod MONOBIO HIC PEG 1000 Å, 5 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152230
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-52230 Supplier Mac-Mod Supplier No. MBHICPEG424610
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)