Learn More
Mac-Mod MONOBIO HILIC Term-Amide 180 Å, 1.8 μm, 2.1 x 100 mm Monodisperse UHPLC Column
SDP
Catalog No. 5000152242
Encompass
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans
Catalog No. 50-001-52242 Supplier Mac-Mod Supplier No. MBHILAMI600210
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans