Learn More
Mac-Mod MONOBIO HILIC Term-Amide 180 Å, 3 μm, 2.1 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000151293
Encompass
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans
Catalog No. 50-001-51293 Supplier Mac-Mod Supplier No. MBHILAMI610210
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans