Learn More
Mac-Mod MONOBIO HILIC Term-Amide 180 Å, 3 μm, 4.6 x 150 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152258
Encompass
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans
Catalog No. 50-001-52258 Supplier Mac-Mod Supplier No. MBHILAMI614615
MONOBIO Term-Amide is a silica based particle with a terminyl amide bonded phase utliized for the analysis of glycans