Learn More
Mac-Mod MONOBIO Pro-IEX SAX, NP, 5 µm, 10.0 x 250 mm Monodisperse HPLC Semi-Prep Column - SS Hardware
SDP
Catalog No. 5000152274
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-52274 Supplier Mac-Mod Supplier No. MBIEXSAXN21025
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations