Learn More
Mac-Mod MONOBIO Pro-IEX SAX NP 5 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152233
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-52233 Supplier Mac-Mod Supplier No. MBIEXSAXN24610
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations