Learn More
Mac-Mod MONOBIO Pro-IEX SCX NP 5 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152283
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-52283 Supplier Mac-Mod Supplier No. MBIEXSCXN24610
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations