Learn More
Mac-Mod MONOBIO Pro-IEX SCX NP 5 µm, 4.6 x 250 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152285
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-52285 Supplier Mac-Mod Supplier No. MBIEXSCXN24625
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations