Learn More
Mac-Mod MONOBIO Pro-IEX WCX NP 5 µm, 4.6 x 50 mm Monodisperse HPLC Column
SDP
Catalog No. 5000152139
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-52139 Supplier Mac-Mod Supplier No. MBIEXWCXN24605
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations