Learn More
Mac-Mod MONOBIO Pro-IEX WCX NP 10 µm, 4.6 x 150 mm Monodisperse HPLC Column
SDP
Catalog No. 5000151870
Encompass
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations
Catalog No. 50-001-51870 Supplier Mac-Mod Supplier No. MBIEXWCXN34615
MONOBIO IEX monodisperse HPLC columns are non-porous particle columns that deliver high efficiency (N) and excellent ion-exchange capacity, to perfrom high resolution separations