Learn More
Mac-Mod MONOBIO RP-DNA C18 1000 Å, 5 µm, 4.6 x 250 mm Monodisperse HPLC Column
SDP
Catalog No. 5000151328
Encompass
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These 1000 Angstrom particle columns provide excellent resoltion and maintain excellent silica structure to ensure long column lifetimes and robust separations.
Catalog No. 50-001-51328 Supplier Mac-Mod Supplier No. MBRPDR18424625D
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These 1000 Angstrom particle columns provide excellent resoltion and maintain excellent silica structure to ensure long column lifetimes and robust separations.