Learn More
Mac-Mod MONOBIO RP-DNA C18 1000 Å, 5 µm, 4.6 x 150 mm Monodisperse HPLC Column
SDP
Catalog No. 5000151964
Encompass
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These 1000 Angstrom particle columns provide excellent resoltion and maintain excellent silica structure to ensure long column lifetimes and robust separations.
Catalog No. 50-001-51964 Supplier Mac-Mod Supplier No. MBRPDR18424615D
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These 1000 Angstrom particle columns provide excellent resoltion and maintain excellent silica structure to ensure long column lifetimes and robust separations.