Learn More
Mac-Mod MONOBIO SEC DIP, 300 Å, 5 µm, 20.0 x 300 mm Monodisperse HPLC Prep Column - SS Hardware
SDP
Catalog No. 5000152029
Encompass
MONOBIO Size-Exclusion monodisperse particle HPLC columns is a high-pore volume particle while maintaining a very narrow particle size distriubtion, which leads to excellent reproducible results. Also, these columns are known for minor shedding of the bonded phase thus being excellent choices for MALS detections. Choose the next evolution in SEC UHPLC/HPLC column analysis!
Catalog No. 50-001-52029 Supplier Mac-Mod Supplier No. MBSECDIP222030
Add to Cart
Add to Cart
Request a Quote
MONOBIO Size-Exclusion monodisperse particle HPLC columns is a high-pore volume particle while maintaining a very narrow particle size distriubtion, which leads to excellent reproducible results. Also, these columns are known for minor shedding of the bonded phase thus being excellent choices for MALS detections. Choose the next evolution in SEC UHPLC/HPLC column analysis!