Learn More
Trajan Scientific HPLC Union Medical Lab 2/PK
SDP
Catalog No. 5000494920
Encompass_Preferred
1/16 to 1/32 ProteCol PEEK unions (2 unions and 4 PEEK ferrules)
Catalog No. 50-004-94920 Supplier Trajan Scientific Supplier No. 212010
1/16 to 1/32 ProteCol PEEK unions (2 unions and 4 PEEK ferrules)