Learn More
Trajan Scientific HPLC Union Medical Lab 2/PK
SDP
Catalog No. 5000494894
Encompass_Preferred
1/32 to 1/32 ProteCol PEEK unions (2 unions and 4 PEEK ferrules)
Catalog No. 50-004-94894 Supplier Trajan Scientific Supplier No. 222009
1/32 to 1/32 ProteCol PEEK unions (2 unions and 4 PEEK ferrules)