Learn More
Foxx Life Sciences EZWaste® HPLC VersaCap® 51S Solvent Waste System, Cap-Only W/ Exhaust Filter, Four (4x) Ports OD Tubing-3.2 mm (1/8"") or 1.6 mm (1/16""), Two (2x) Ports OD Tubing 6.4mm (1/4""), 1/EA
GREENER_CHOICE
Catalog No. 501720602
Encompass_Preferred
Foxx Life Sciences EZWaste® HPLC VersaCap® 51S Solvent Waste Cap Assembly is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from multiple HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides safer mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.Each EZwaste® Assembly includes a VersaCap® 51S with Manifold, fittings 4 x 3.2 mm (1/8") OD Tubing, fittings 4 x 1.6 mm (1/16") OD Tubing, fittings 2 x 6.4mm (1/4) OD Tubing , All Plugs suitable for blocking of unused ports & 1 carbon exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary.
Catalog No. 50-172-0602 Supplier Foxx Life Sciences Supplier No. 3300A51FLS
Foxx Life Sciences EZWaste® HPLC VersaCap® 51S Solvent Waste Cap Assembly is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from multiple HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides safer mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.Each EZwaste® Assembly includes a VersaCap® 51S with Manifold, fittings 4 x 3.2 mm (1/8") OD Tubing, fittings 4 x 1.6 mm (1/16") OD Tubing, fittings 2 x 6.4mm (1/4) OD Tubing , All Plugs suitable for blocking of unused ports & 1 carbon exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary.