Learn More
Foxx Life Sciences EZWaste® HPLC Universal Stackable VersaCap® 53mm(53B) Solvent Waste System, Cap-Only W/Exhaust Filter, Six(6x) Ports OD Tubing -3.2 mm (1/8"") or 1.6 mm(1/16""), Three(3x) Hose Barb 3.2mm (1/8"") or 9.5mm(3/8""), Three(3x) OD Tubing 12.7 mm (1/2""), 1/EA
GREENER_CHOICE
Catalog No. 501720606
Encompass_Preferred
Foxx Life Sciences EZwaste® Universal Stackable HPLC VersaCap® GL45 Solvent Waste System is designed to be an affordable solution for HPLC liquid waste collection & disposal. System features an activated carbon filter which traps volatile organic compounds from being released into the air. The EZwaste® HPLC Solvent Waste system also features a closed loop design which stops leaks & improves overall lab safety. In EZwaste® Universal Stackable design you can add multiple manifolds for increased ports, conveniently as per requirement.Each EZwaste® Assy. includes a VersaCap® GL45, Universal Stackable Manifold w/ 6-3-3 Ports, 6x fittings of 3.2mm (1/8") OD Tubing, 6x fittings of 1.6 mm (1/16") OD Tubing, 3x fittings of 3.2mm (1/8") Hose Barb, 3x fittings of 9.5mm (3/8") Hose Barb, 3x fittings of 12.7mm (1/2") OD Tubing , 1 carbon Exhaust Filter & all suitable plugs/adapters for blocking of unused ports. * Carbon exhaust filter life may very.
Catalog No. 50-172-0606 Supplier Foxx Life Sciences Supplier No. 3300214FLS
Foxx Life Sciences EZwaste® Universal Stackable HPLC VersaCap® GL45 Solvent Waste System is designed to be an affordable solution for HPLC liquid waste collection & disposal. System features an activated carbon filter which traps volatile organic compounds from being released into the air. The EZwaste® HPLC Solvent Waste system also features a closed loop design which stops leaks & improves overall lab safety. In EZwaste® Universal Stackable design you can add multiple manifolds for increased ports, conveniently as per requirement.Each EZwaste® Assy. includes a VersaCap® GL45, Universal Stackable Manifold w/ 6-3-3 Ports, 6x fittings of 3.2mm (1/8") OD Tubing, 6x fittings of 1.6 mm (1/16") OD Tubing, 3x fittings of 3.2mm (1/8") Hose Barb, 3x fittings of 9.5mm (3/8") Hose Barb, 3x fittings of 12.7mm (1/2") OD Tubing , 1 carbon Exhaust Filter & all suitable plugs/adapters for blocking of unused ports. * Carbon exhaust filter life may very.