Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, 20L Heavy Duty Reusable Carboy, 83mm (83B) VersaCap with Four (4x) Ports for 1/16 Inch O.D. Tubing; One (1x) Port for 1/4 Inch O.D. Tubing; Three (3x) Hose Barb Ports for 1/4 or 3/8 Inch I.D. Tubing
GREENER_CHOICE
Catalog No. 501178600
Encompass_Preferred
332-4426-OEM: EZwaste System, 20L, HDPE, 83mm Cap, 4x 1/16, 1x 1/4 OD Tube Fittings, 3 HB & Filter. Foxx Life Sciences EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design, which protects against leaks and spills, improving overall lab safety. Each EZwaste Solvent Waste System comes with one 83mm or 120mm open VersaCap, fittings, plugs for unused ports, a replaceable sulfur-cured EPDM gasket, a chemical exhaust filter, and a chemically resilient container. *Depending on application, carbon exhaust filter life cycle may vary.
Catalog No. 50-117-8600 Supplier Foxx Life Sciences Supplier No. 3324426OEM
332-4426-OEM: EZwaste System, 20L, HDPE, 83mm Cap, 4x 1/16, 1x 1/4 OD Tube Fittings, 3 HB & Filter. Foxx Life Sciences EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design, which protects against leaks and spills, improving overall lab safety. Each EZwaste Solvent Waste System comes with one 83mm or 120mm open VersaCap, fittings, plugs for unused ports, a replaceable sulfur-cured EPDM gasket, a chemical exhaust filter, and a chemically resilient container. *Depending on application, carbon exhaust filter life cycle may vary.