Learn More
Agilent Technologies InfinityLab Screw Plug, PTFE, 1/8 in.-27 NPT Thread, 1/Pk.
Catalog No. NC3627455
Encompass
This screw plug is designed for use as a leak hose port component in liquid chromatography (LC) applications. Constructed from PTFE, it provides high chemical resistance and is intended for use with pump modules in analytical systems.
  • Constructed from durable polytetrafluorethylene (PTFE)
  • Features 1/8"-27 NPT threading
  • Designed for analytical LC pump module compatibility
  • Includes one unit per package
Catalog No. NC3627455 Supplier Agilent Technologies Supplier No. 50431195
This screw plug is designed for use as a leak hose port component in liquid chromatography (LC) applications. Constructed from PTFE, it provides high chemical resistance and is intended for use with pump modules in analytical systems.
  • Constructed from durable polytetrafluorethylene (PTFE)
  • Features 1/8"-27 NPT threading
  • Designed for analytical LC pump module compatibility
  • Includes one unit per package