Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, Cap-Only Assembly w/ Drum Bung Adapter, 70mm (S70) VersaCap w/ Four (4x) Ports for 1/8 Inch O.D. Tubing; One (1x) Port for 1/4 Inch O.D. Tubing; Three (3x) Hose Barb Ports for 1/4 or 3/8 Inch I.D. Tubing
GREENER_CHOICE
Catalog No. 501178562
Encompass_Preferred
330-0C19-OEM: EZwaste System, UN/DOT, 70mm Cap w/ Threaded Adapter, 4x 1/8, 1x 1/4 OD Tube Fittings, 3 HB & Filter. Foxx Life Sciences EZwaste Solvent Waste System is designed to be an affordable solution for HPLC waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds. This prevents chemicals from being inhaled by lab workers, & provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design, which protects against leaks & spills, improving overall lab safety. Each EZwaste Solvent Waste System comes with one 83mm or 120mm open VersaCap, fittings, plugs for unused ports, a replaceable sulfur-cured EPDM gasket, & a chemical exhaust filter. This item also includes a threaded bung adapter for connection with large drums. *Depending on application, carbon exhaust filter life cycle may vary.
Catalog No. 50-117-8562 Supplier Foxx Life Sciences Supplier No. 3300C19OEM
330-0C19-OEM: EZwaste System, UN/DOT, 70mm Cap w/ Threaded Adapter, 4x 1/8, 1x 1/4 OD Tube Fittings, 3 HB & Filter. Foxx Life Sciences EZwaste Solvent Waste System is designed to be an affordable solution for HPLC waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds. This prevents chemicals from being inhaled by lab workers, & provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design, which protects against leaks & spills, improving overall lab safety. Each EZwaste Solvent Waste System comes with one 83mm or 120mm open VersaCap, fittings, plugs for unused ports, a replaceable sulfur-cured EPDM gasket, & a chemical exhaust filter. This item also includes a threaded bung adapter for connection with large drums. *Depending on application, carbon exhaust filter life cycle may vary.