Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPLC Water, MilliporeSigma™ PROMO

Catalog No. MWX00081S24 Shop All MilliporeSigma Products
Change view
Click to view available options
HPLC Water; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MWX00081S24 HPLC Water; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWX00081S24 Supplier MilliporeSigma Supplier No. MWX00081S24

Synonym: Dihydrogen Monoxide, Dihydrogen Oxide, Distilled Water, Sterile Water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name water
SMILES O

Specifications

Absorbance 0.005 AU at 300 to 400 nm
Color Colorless
Boiling Point 100°C
Linear Formula H2O
Formula Weight 18.2
Grade HPLC
Residue after Evaporation 2 ppm Maximum
Vapor Pressure 23 hPa (20°C)
Quantity 4 L
Refractive Index 1.332 to 1.334
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
RTECSNumber ZC0110000
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.