Learn More
Foxx Life Sciences PUREGRIP® HPLC Solvent Reservoir System with VersaCap® GL45 and 500mL Amber Glass Reagent Bottle, Class VI PTFE Adapter & Safety Air Inlet Valve, Three (3x) Ports- OD Tubing 3.2mm (1/8"") or 1.6mm (1/16""), 1/EA
Catalog No. 501720634
Encompass_Preferred
Foxx Life Sciences PUREGRIP® HPLC Solvent Reservoir Caps & Bottle system offer the safest & most ergonomic lab safety solution in the industry. This system come w/ VersaCap®, a Class VI PTFE Manifold, chemically resistant Viton O-Ring & Air Inlet Valve combined w/ a Foxx 25mm, 0.2m PTFE Filter for safe vapor filtration.The Air Inlet Valve & PTFE Filter block out all harmful solvent vapors while blocking any incoming contaminants from the incoming air.The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap & eliminates the tubes from sliding in & out of the container.Each assembly comes w/ VersaCap® GL45 Cap & Manifold & Bottle Assy., Safety Inlet vale w/ filter, fittings for 3x 3.2mm (1/8") OD Tubing, 3x 1.6mm (1/16") OD Tubing & 3x Suitable Plugs blocking of unused ports.*Filter absorbs contaminants from the flowing air, it is recommended that one replaces the vent filter every 6 months to ensure quality operation.
Catalog No. 50-172-0634 Supplier Foxx Life Sciences Supplier No. 45V4602FLS
Foxx Life Sciences PUREGRIP® HPLC Solvent Reservoir Caps & Bottle system offer the safest & most ergonomic lab safety solution in the industry. This system come w/ VersaCap®, a Class VI PTFE Manifold, chemically resistant Viton O-Ring & Air Inlet Valve combined w/ a Foxx 25mm, 0.2m PTFE Filter for safe vapor filtration.The Air Inlet Valve & PTFE Filter block out all harmful solvent vapors while blocking any incoming contaminants from the incoming air.The sealed VersaCap® design that Foxx offers eliminates tubes twisting as you thread the cap & eliminates the tubes from sliding in & out of the container.Each assembly comes w/ VersaCap® GL45 Cap & Manifold & Bottle Assy., Safety Inlet vale w/ filter, fittings for 3x 3.2mm (1/8") OD Tubing, 3x 1.6mm (1/16") OD Tubing & 3x Suitable Plugs blocking of unused ports.*Filter absorbs contaminants from the flowing air, it is recommended that one replaces the vent filter every 6 months to ensure quality operation.