Learn More
Mac-Mod Avantor Alltima C18, 100 Å, 5 µm, 4.6 x 250 mm HPLC Column-MF (PEEK)
SDP
Catalog No. 5000143443
Encompass
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-001-43443 Supplier Mac-Mod Supplier No. 88055 / 5129677
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.