Learn More
Mac-Mod Avantor Alltima Silica, 100 Å, 5 µm, 4.6 x 250 mm HPLC Column
SDP
Catalog No. 5000143151
Encompass
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-001-43151 Supplier Mac-Mod Supplier No. 88171 / 5129707
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.