Learn More
Mac-Mod Avantor Apex ODS, 100 Å, 5 µm, 4.6 x 100 mm HPLC Column
SDP
Catalog No. 5000143386
Encompass
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 Å pore size spherical silica particle column. Narrow particle size distribution.
Catalog No. 50-001-43386 Supplier Mac-Mod Supplier No. 4M10310 / 5109697
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 Å pore size spherical silica particle column. Narrow particle size distribution.