Learn More
Mac-Mod Avantor ACE - Bioanalytical C18-300, 300 Å, 3 µm, 3.0 x 150 mm HPLC Column
SDP
Supplier:  Mac-Mod ACE2111503
Encompass
Avantor ACE Bioanalytical HPLC columns are manufactured with 300 A Ultra-Inert Base Deactivated Silica specifically designed for your Protein and Peptide HPLC method development needs. Avantor ACE Bioanalytical HPLC columns are available in a wide range of bonded phases (including C18, C8, C4, Phenyl, and CN). Avantor ACE Bioanalytical HPLC columns are available in 3, and 5 um particle sizes and a wide range of column IDs and column lengths including nano, capillary, microbore, analytical, semi-preparative and preparative formats. All Avantor ACE Bioanalytical columns are known to deliver excellent peak shape and verified batch to batch reproducibility for difficult Protein and Peptide separations. Columns are packed in 275 bar HPLC hardware. Note: Avantor ACE Bioanalytical HPLC guard columns are available for all 3.0 and 5.0 um particle sizes across 2.1 mm to 4.6 mm column ID - C0001 male/male union is needed to connect guard holder (H0001) to the analytical column.
Catalog No. 50-001-43346