Learn More
Mac-Mod Avantor ACE UltraCore SuperC18, 95 Å, 2.5 µm, 0.5 x 150 mm Solid-Core Column
SDP
Catalog No. 5000144197
Encompass
Avantor ACE UltraCore solid-core (core-shell) particle columns are ideal for MS work offering improved signal intensity and maximum response. These columns combine high efficiency separations with low back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability. Solid Core particle technology provides higher efficiencies per unit back pressure as compared to sub-2-micron UHPLC columns. Columns combine the proven advantages of UHPLC and HPLC columns with Solid Core Particle Technology. Solid-core columns can be used on both UHPLC and HPLC systems. Excellent peak shape and phases stability with extended pH. Highly inert, ultra-low bleed, designed for use with LC/MS compatible buffers. 2.5 and 5 µm particle sizes available.
Catalog No. 50-001-44197 Supplier Mac-Mod Supplier No. CORE25A15005
Avantor ACE UltraCore solid-core (core-shell) particle columns are ideal for MS work offering improved signal intensity and maximum response. These columns combine high efficiency separations with low back pressure. The Encapsulated Bonding Technology (EBT™) greatly increases the ligand coverage of the silica surface and eliminates the effect of unbonded silanol groups on separations. This higher ligand coverage results in improved inertness, chromatographic performance and stability. Solid Core particle technology provides higher efficiencies per unit back pressure as compared to sub-2-micron UHPLC columns. Columns combine the proven advantages of UHPLC and HPLC columns with Solid Core Particle Technology. Solid-core columns can be used on both UHPLC and HPLC systems. Excellent peak shape and phases stability with extended pH. Highly inert, ultra-low bleed, designed for use with LC/MS compatible buffers. 2.5 and 5 µm particle sizes available.