Learn More
Mac-Mod Avantor Alltima Phenyl, 100 Å, 3 µm, 4.6 x 50 mm HPLC Column
SDP
Catalog No. 5000144015
Encompass
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-001-44015 Supplier Mac-Mod Supplier No. 81178
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.