Learn More
Mac-Mod Avantor Genesis C8, 120 Å, 4 µm, 2.1 x 50 mm HPLC Column
SDP
Catalog No. 5000143878
Encompass
Avantor Genesis phases are based on high purity metal-free spherical silica.
Catalog No. 50-001-43878 Supplier Mac-Mod Supplier No. FK5962E / 5109766
Avantor Genesis phases are based on high purity metal-free spherical silica.